Thermo Scientific Chemicals

DL-Thioctic Acid, 98+%

Catalog number: 138720050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-Thioctic Acid, 98+%

Catalog number: 138720050
5 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 138720050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1077-28-7
IUPAC Name
5-(1,2-dithiolan-3-yl)pentanoic acid
Molecular Formula
C8H14O2S2
InChI Key
AGBQKNBQESQNJD-UHFFFAOYNA-N
SMILES
OC(=O)CCCCC1CCSS1
UV spectrum
Complies
Melting point
60°C to 63°C
UV spectrophotometry
98.0 to 102.0 % (on dry substance)
Sulfated ash
=<0.1 %
Solubility
(2 % in 10 % NaOH) Clear yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text