Thermo Scientific Chemicals

2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals

Catalog number: 138790025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals

Catalog number: 138790025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 138790025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-42-5
IUPAC Name
2-sulfanylpropanoic acid amine
Molecular Formula
C3H9NO2S
InChI Key
VHFSMDHANDABQK-UHFFFAOYNA-N
SMILES
N.CC(S)C(O)=O
Refractive index
1.4804 to 1.4824 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Titration Iodimetric
>=94 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text