Thermo Scientific Chemicals

Thionicotinamide, 99%, Thermo Scientific Chemicals

Catalog number: 138800250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Thionicotinamide, 99%, Thermo Scientific Chemicals

Catalog number: 138800250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 138800250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4621-66-3
IUPAC Name
pyridine-3-carbothioamide
Molecular Formula
C6H6N2S
InChI Key
XQWBMZWDJAZPPX-UHFFFAOYSA-N
SMILES
NC(=S)C1=CC=CN=C1
Appearance (Form)
Powder
Melting point
183°C to 190°C (Dec)
Appearance (Color)
Yellow to yellow-green
Infrared spectrum
Conforms
Titration with HClO4
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text