Thionicotinamide, 99%, Thermo Scientific Chemicals
Thionicotinamide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thionicotinamide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 138801000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4621-66-3
IUPAC Namepyridine-3-carbothioamide
Molecular FormulaC6H6N2S
InChI KeyXQWBMZWDJAZPPX-UHFFFAOYSA-N
SMILESNC(=S)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Appearance (Form)Powder
Melting point183°C to 190°C (Dec)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only