Thermo Scientific Chemicals

2-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: 138840250
25 g, Each
Thermo Scientific Chemicals

2-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: 138840250
25 g, Each
Quantity
Catalog number: 138840250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
527-72-0
IUPAC Name
thiophene-2-carboxylic acid
Molecular Formula
C5H4O2S
InChI Key
QERYCTSHXKAMIS-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CS1
Appearance (Color)
White to cream to light brown
Appearance (Form)
Crystalline powder
Melting point
126°C to 130°C
Titration with NaOH
98.5 to 101.5 %
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text