2-Thiophenecarboxylic acid, 99%
2-Thiophenecarboxylic acid, 99%
2-Thiophenecarboxylic acid, 99%
2-Thiophenecarboxylic acid, 99%
Thermo Scientific Chemicals

2-Thiophenecarboxylic acid, 99%

CAS: 527-72-0 | C5H4O2S | 128.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 138845000
Price (EUR)/ Each
146,20
Online Exclusive
172,00
Save 25,80 (15%)
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)/ Each
146,20
Online Exclusive
172,00
Save 25,80 (15%)
Add to cart
Chemical Identifiers
IUPAC Namethiophene-2-carboxylic acid
Molecular FormulaC5H4O2S
InChI KeyQERYCTSHXKAMIS-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CS1
Molecular Weight (g/mol)128.15
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to cream to light brown
Melting point126°C to 130°C
GC>=98.5 %
Titration with NaOH98.5 to 101.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link