Thiosemicarbazide, 98+%, Thermo Scientific Chemicals
Thiosemicarbazide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thiosemicarbazide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number 138905000
Price (USD)/ Each
86.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
102.00 
Save 15.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
86.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
102.00 
Save 15.35 (15%)
Add to cart
Thiosemicarbazide, 98+%, Thermo Scientific Chemicals
Catalog number138905000
Price (USD)/ Each
86.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
102.00 
Save 15.35 (15%)
-
Add to cart
Chemical Identifiers
CAS79-19-6
IUPAC Nameaminothiourea
Molecular FormulaCH5N3S
InChI KeyBRWIZMBXBAOCCF-UHFFFAOYSA-N
SMILESNNC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point175°C to 180°C
HPLC>=98.0 %
Appearance (Color)White to yellow to beige
Appearance (Form)Powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only