Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thiourea, 99%, extra pure, Thermo Scientific Chemicals

Thiourea, Thiocarbamide, CAS # 62-56-6, is an analogue of urea with a single sulfur-substituted oxygen.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 138910050
Price (USD)/ Each
277.00
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
277.00
Add to cart
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Catalog number138910050
Price (USD)/ Each
277.00
-
Add to cart
Chemical Identifiers
CAS62-56-6
IUPAC Namethiourea
Molecular FormulaCH4N2S
InChI KeyUMGDCJDMYOKAJW-UHFFFAOYSA-N
SMILESNC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point171°C to 177°C
Titration Iodimetric>=98.5 %
Loss on drying=<0.5 %
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Thiourea is a derivative of the biological compound, urea
• Thiourea is highly toxic when it decomposes and acts as a carcinogen

Applications

• Thiourea can be used as a denaturant in lysis buffers and similar sample buffers that require solubilization of proteins
• Thiourea can act as an antioxidant and chromophore
• Thiourea can be used in the study of cellular redox, autophagy, and aging

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Wang, Y.; Jin, X.; He, L.; Zhang, W. Inhibitory effect of thiourea on biological nitrification process and its eliminating method. Water Sci Technol. 2017 75(12), 2900-2907.
  2. Park, S.; Lee, H.; Chae, J.B.; Kim, C. A Thiourea-Containing Fluorescent Chemosensor for Detecting Ga3. J Fluoresc. 2020 30(6), 1457-1462.