Thermo Scientific Chemicals

L-Threonine, 98%

Catalog number: 138931000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

L-Threonine, 98%

Catalog number: 138931000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 138931000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
72-19-5
IUPAC Name
2-amino-3-hydroxybutanoic acid
Molecular Formula
C4H9NO3
InChI Key
AYFVYJQAPQTCCC-UHFFFAOYNA-N
SMILES
CC(O)C(N)C(O)=O
Appearance (Form)
Crystalline powder or glistening flakes
Titration with HClO4
97.5 to 102.5 % (On dried substance)
Heavy metals (as Pb)
=<10 ppm
Chloride (Cl)
=<200 ppm
Sulfate (SO4)
=<300 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text