DL-Threonine, 99.5%
Thermo Scientific Chemicals

DL-Threonine, 99.5%

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number 138945000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS80-68-2
IUPAC Name(2R,3R)-2-amino-3-hydroxybutanoic acid
Molecular FormulaC4H9NO3
InChI KeyAYFVYJQAPQTCCC-PWNYCUMCSA-N
SMILESC[C@@H](O)[C@@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Titration with HClO498.0 to 101.0 % (On dried substance)
Loss on drying=<0.2 % (1 g, 105°C, 3 hrs)
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only