Thermo Scientific Chemicals

trans-2,3-Dimethylacrylic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 138970250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

trans-2,3-Dimethylacrylic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 138970250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 138970250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
80-59-1
IUPAC Name
(2E)-2-methylbut-2-enoic acid
Molecular Formula
C5H8O2
InChI Key
UIERETOOQGIECD-ONEGZZNKSA-N
SMILES
C\C=C(/C)C(O)=O
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %
Appearance (Color)
White to beige
Appearance (Form)
Crystalline powder and/or chunks
Melting point
61°C to 65°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text