trans-2,3-Dimethylacrylic Acid, 98%, Thermo Scientific Chemicals
trans-2,3-Dimethylacrylic Acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-2,3-Dimethylacrylic Acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 138975000
Price (USD)/ Each
320.00
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
320.00
Add to cart
trans-2,3-Dimethylacrylic Acid, 98%, Thermo Scientific Chemicals
Catalog number138975000
Price (USD)/ Each
320.00
-
Add to cart
Chemical Identifiers
CAS80-59-1
IUPAC Name(2E)-2-methylbut-2-enoic acid
Molecular FormulaC5H8O2
InChI KeyUIERETOOQGIECD-ONEGZZNKSA-N
SMILESC\C=C(/C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Color)White to beige
Appearance (Form)Crystalline powder and/or chunks
Melting point61°C to 65°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only