Tolazoline hydrochloride, 99%
Tolazoline hydrochloride, 99%
Thermo Scientific Chemicals

Tolazoline hydrochloride, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number 138980500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS59-97-2
IUPAC Namehydrogen 2-benzyl-4,5-dihydro-1H-imidazole chloride
Molecular FormulaC10H13ClN2
InChI KeyRHTNTTODYGNRSP-UHFFFAOYSA-N
SMILES[H+].[Cl-].C(C1=NCCN1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow
Melting point172°C to 176°C
Titration with HClO4>=98.5 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only