Thermo Scientific Chemicals

Tolazoline hydrochloride, 99%

Catalog number: 138982500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Tolazoline hydrochloride, 99%

Catalog number: 138982500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 138982500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59-97-2
IUPAC Name
hydrogen 2-benzyl-4,5-dihydro-1H-imidazole chloride
Molecular Formula
C10H13ClN2
InChI Key
RHTNTTODYGNRSP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].C(C1=NCCN1)C1=CC=CC=C1
Melting point
172°C to 176°C
Titration with HClO4
>=98.5 %
Infrared spectrum
Conforms
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text