Thermo Scientific Chemicals

m-Tolualdehyde, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 138990250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

m-Tolualdehyde, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 138990250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 138990250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
620-23-5
IUPAC Name
3-methylbenzaldehyde
Molecular Formula
C8H8O
InChI Key
OVWYEQOVUDKZNU-UHFFFAOYSA-N
SMILES
CC1=CC=CC(C=O)=C1
GC
>=97.5 %
Stabilizer
0.1 % hydroquinone
Appearance (Color)
Clear yellow to brown
Appearance
May darken on storage
Refractive index
1.5400 to 1.5420 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text