m-Tolualdehyde, 98%, stabilized, Thermo Scientific Chemicals
m-Tolualdehyde, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

m-Tolualdehyde, 98%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 138991000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS620-23-5
IUPAC Name3-methylbenzaldehyde
Molecular FormulaC8H8O
InChI KeyOVWYEQOVUDKZNU-UHFFFAOYSA-N
SMILESCC1=CC=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
AppearanceMay darken on storage
Refractive index1.5400 to 1.5420 (20°C, 589 nm)
GC>=97.5 %
Stabilizer0.1 % hydroquinone
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only