Thermo Scientific Chemicals

p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 139015000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 139015000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 139015000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207801-20-5
IUPAC Name
sodium 4-methylbenzene-1-sulfinate
Molecular Formula
C7H7NaO2S
InChI Key
KFZUDNZQQCWGKF-UHFFFAOYSA-M
SMILES
[Na+].CC1=CC=C(C=C1)S([O-])=O
Appearance (Form)
Crystalline powder or flakes
Water
=<5 % (K.F.)
Titration with HClO4
>=98.0 % (On dry substance)
Infrared spectrum
Conforms
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text