p-Toluenesulfonyl chloride, 99+%
p-Toluenesulfonyl chloride, 99+%
p-Toluenesulfonyl chloride, 99+%
p-Toluenesulfonyl chloride, 99+%
Thermo Scientific Chemicals

p-Toluenesulfonyl chloride, 99+%

CAS: 98-59-9 | C7H7ClO2S | 190.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 139030250
Price (EUR)
20,91
Online Exclusive
24,60
Save 3,69 (15%)
Each
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
20,91
Online Exclusive
24,60
Save 3,69 (15%)
Each
Add to cart
Chemical Identifiers
CAS98-59-9
IUPAC Name4-methylbenzene-1-sulfonyl chloride
Molecular FormulaC7H7ClO2S
InChI KeyYYROPELSRYBVMQ-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White
Infrared spectrumConforms
Melting point67°C to 69°C
Assay>=99.0 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469183Certificate of AnalysisJan 31, 2025139030250, 139030100, 139035000, 139030020, 139031000
A0469184Certificate of AnalysisJan 31, 2025139030250, 139030100, 139035000, 139030020, 139031000
A0464119Certificate of AnalysisJun 27, 2024139030250, 139030100, 139035000, 139030020, 139031000
A0455369Certificate of AnalysisAug 25, 2023139030250, 139030100, 139035000, 139030020, 139031000
A0402054Certificate of AnalysisDec 15, 2022139030250, 139030100, 139035000, 139030020, 139031000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Substances/mixtures corrosive to metal
Category 1
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 1
Skin Sensitization
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H315 - Causes skin irritation

H317 - May cause an allergic skin reaction

H318 - Causes serious eye damage

Physical hazards

H290 - May be corrosive to metals

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

P402 + P404 - Store in a dry place. Store in a closed container

Share catalog number, name or link

1x1 image pixel for data collection