Thermo Scientific Chemicals

p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals

Catalog number: 139035000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals

Catalog number: 139035000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 139035000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-59-9
IUPAC Name
4-methylbenzene-1-sulfonyl chloride
Molecular Formula
C7H7ClO2S
InChI Key
YYROPELSRYBVMQ-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)S(Cl)(=O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
67°C to 69°C
Assay
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text