o-Tolylacetic acid, 99%, Thermo Scientific Chemicals
o-Tolylacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

o-Tolylacetic acid, 99%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
25 g
Packaging:
Glass Bottle
Catalog number 139100250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS644-36-0
IUPAC Name2-(2-methylphenyl)acetic acid
Molecular FormulaC9H10O2
InChI KeyRZWGTXHSYZGXKF-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
HPLC>=98.5 %
Melting point87°C to 90°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only