Thermo Scientific Chemicals

m-Tolylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 139112500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

m-Tolylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 139112500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 139112500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
621-36-3
IUPAC Name
2-(3-methylphenyl)acetic acid
Molecular Formula
C9H10O2
InChI Key
GJMPSRSMBJLKKB-UHFFFAOYSA-N
SMILES
CC1=CC=CC(CC(O)=O)=C1
Appearance (Form)
Glistening flakes and/or chunks
Melting point
61°C to 66°C
Appearance (Color)
Colorless to off-white
Infrared spectrum
Conforms
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text