Thermo Scientific Chemicals

p-Tolylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: 139120250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

p-Tolylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: 139120250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 139120250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
622-47-9
IUPAC Name
2-(4-methylphenyl)acetate
Molecular Formula
C9H9O2
InChI Key
GXXXUZIRGXYDFP-UHFFFAOYSA-M
SMILES
CC1=CC=C(CC([O-])=O)C=C1
Titration with NaOH
>=98.5 %
Appearance (Form)
Crystalline powder
Melting point
90°C to 93°C
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text