Chemical Identifiers
IUPAC Namehydrogen methyl 5-[(diaminomethylidene)amino]-2-(4-methylbenzenesulfonamido)pentanoate chloride
Molecular FormulaC14H23ClN4O4S
InChI KeyJIQFFACVQXXHMY-UHFFFAOYNA-N
SMILES[H+].[Cl-].COC(=O)C(CCCN=C(N)N)NS(=O)(=O)C1=CC=C(C)C=C1
Molecular Weight (g/mol)378.87
View more