Thermo Scientific Chemicals

Nalpha-4-Tosyl-L-arginine methyl ester hydrochloride, 98+%

Catalog number: 139200500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Nalpha-4-Tosyl-L-arginine methyl ester hydrochloride, 98+%

Catalog number: 139200500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 139200500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1784-03-8
IUPAC Name
hydrogen methyl 5-[(diaminomethylidene)amino]-2-(4-methylbenzenesulfonamido)pentanoate chloride
Molecular Formula
C14H23ClN4O4S
InChI Key
JIQFFACVQXXHMY-UHFFFAOYNA-N
SMILES
[H+].[Cl-].COC(=O)C(CCCN=C(N)N)NS(=O)(=O)C1=CC=C(C)C=C1
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Melting point
146°C to 149°C
Infrared spectrum
Conforms
Titration with NaOH
98.0 to 102.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text