Thermo Scientific Chemicals

Triacetin, 99%, Thermo Scientific Chemicals

Catalog number: 139220025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Triacetin, 99%, Thermo Scientific Chemicals

Catalog number: 139220025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 139220025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
102-76-1
IUPAC Name
1,3-bis(acetyloxy)propan-2-yl acetate
Molecular Formula
C9H14O6
InChI Key
URAYPUMNDPQOKB-UHFFFAOYSA-N
SMILES
CC(=O)OCC(COC(C)=O)OC(C)=O
Refractive index
1.4290 to 1.4330 (20°C, 589 nm)
Appearance (Form)
Liquid
Density
(25°C) 1.154 to 1.158 g/ml
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text