Triacetin, 99%
Triacetin, 99%
Triacetin, 99%
Thermo Scientific Chemicals

Triacetin, 99%

CAS: 102-76-1 | C9H14O6 | 218.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
10 L
500 mL
1 L
Packaging:
View all
Glass bottle
Catalog number 139225000
Price (EUR)
33,49
Online Exclusive
39,40
Save 5,91 (15%)
Each
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
33,49
Online Exclusive
39,40
Save 5,91 (15%)
Each
Add to cart
Chemical Identifiers
CAS102-76-1
IUPAC Name1,3-bis(acetyloxy)propan-2-yl acetate
Molecular FormulaC9H14O6
InChI KeyURAYPUMNDPQOKB-UHFFFAOYSA-N
SMILESCC(=O)OCC(COC(C)=O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4290 to 1.4330 (20°C, 589 nm)
Density(25°C) 1.154 to 1.158 g/ml
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link