Thermo Scientific Chemicals

2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: 139250250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: 139250250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 139250250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1004-38-2
IUPAC Name
pyrimidine-2,4,6-triamine
Molecular Formula
C4H7N5
InChI Key
JTTIOYHBNXDJOD-UHFFFAOYSA-N
SMILES
NC1=CC(N)=NC(N)=N1
Appearance (Color)
Almost white to yellow to light brown
Titration with HClO4
96.0 to 104.0 %
Infrared spectrum
Conforms
Appearance (Form)
Powder
Melting point
246°C to 252°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text