2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 139251000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namepyrimidine-2,4,6-triamine
Molecular FormulaC4H7N5
InChI KeyJTTIOYHBNXDJOD-UHFFFAOYSA-N
SMILESNC1=CC(N)=NC(N)=N1
Molecular Weight (g/mol)125.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point246°C to 252°C
Appearance (Color)Almost white to yellow to light brown
Titration with HClO496.0 to 104.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only