Thermo Scientific Chemicals

1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals

Catalog number: 139285000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals

Catalog number: 139285000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 139285000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
288-88-0
IUPAC Name
1H-1,2,4-triazole
Molecular Formula
C2H3N3
InChI Key
NSPMIYGKQJPBQR-UHFFFAOYSA-N
SMILES
N1C=NC=N1
Appearance (Form)
Powder and/or flakes
Infrared spectrum
Conforms
Melting point
119°C to 122°C
Appearance (Color)
White
HPLC
>=99.4 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text