Tribromoacetic acid, 99%, Thermo Scientific Chemicals
Tribromoacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tribromoacetic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
Catalog number 139300250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS75-96-7
IUPAC Nametribromoacetate
Molecular FormulaC2Br3O2
InChI KeyQIONYIKHPASLHO-UHFFFAOYSA-M
SMILES[O-]C(=O)C(Br)(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystals
Infrared spectrumConforms
Melting point130°C to 133°C
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only