Thermo Scientific Chemicals

Tributylamine, 99%, Thermo Scientific Chemicals

Catalog number: 139320025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Tributylamine, 99%, Thermo Scientific Chemicals

Catalog number: 139320025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 139320025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
102-82-9
IUPAC Name
tributylamine
Molecular Formula
C12H27N
InChI Key
IMFACGCPASFAPR-UHFFFAOYSA-N
SMILES
CCCCN(CCCC)CCCC
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.4273 to 1.4293 (20°C, 589 nm)
Appearance (Form)
Clear liquid
Color scale
=<20 APHA

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text