Tri-n-butylphosphine, 95%, Thermo Scientific Chemicals
Tri-n-butylphosphine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-n-butylphosphine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 139341000
Price (USD)/ Each
59.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
70.10 
Save 10.45 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
59.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
70.10 
Save 10.45 (15%)
Add to cart
Tri-n-butylphosphine, 95%, Thermo Scientific Chemicals
Catalog number139341000
Price (USD)/ Each
59.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
70.10 
Save 10.45 (15%)
-
Add to cart
Chemical Identifiers
CAS998-40-3
IUPAC Nametributylphosphane
Molecular FormulaC12H27P
InChI KeyTUQOTMZNTHZOKS-UHFFFAOYSA-N
SMILESCCCCP(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4610 to 1.4630 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only