Tricarballylic acid, 99%, Thermo Scientific Chemicals
Tricarballylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tricarballylic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number 139360050
Price (USD)/ Each
38.65
Online exclusive
42.50 
Save 3.85 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
38.65
Online exclusive
42.50 
Save 3.85 (9%)
Add to cart
Tricarballylic acid, 99%, Thermo Scientific Chemicals
Catalog number139360050
Price (USD)/ Each
38.65
Online exclusive
42.50 
Save 3.85 (9%)
-
Add to cart
Chemical Identifiers
CAS99-14-9
IUPAC Namepropane-1,2,3-tricarboxylic acid
Molecular FormulaC6H8O6
InChI KeyKQTIIICEAUMSDG-UHFFFAOYSA-N
SMILESOC(=O)CC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point154°C to 159°C
Titration with TBAH>=98.5 %
Appearance (Color)Off-white to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only