Trichloroacetonitrile, 98%, Thermo Scientific Chemicals
Trichloroacetonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trichloroacetonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
Glass bottle
Catalog number 139371000
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
Add to cart
Trichloroacetonitrile, 98%, Thermo Scientific Chemicals
Catalog number139371000
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS545-06-2
IUPAC Nametrichloroacetonitrile
Molecular FormulaC2Cl3N
InChI KeyDRUIESSIVFYOMK-UHFFFAOYSA-N
SMILESClC(Cl)(Cl)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4400 to 1.4420 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only