Thermo Scientific Chemicals

Trichloroacetonitrile, 98%, Thermo Scientific Chemicals

Catalog number: 139375000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Trichloroacetonitrile, 98%, Thermo Scientific Chemicals

Catalog number: 139375000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 139375000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
545-06-2
IUPAC Name
trichloroacetonitrile
Molecular Formula
C2Cl3N
InChI Key
DRUIESSIVFYOMK-UHFFFAOYSA-N
SMILES
ClC(Cl)(Cl)C#N
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.4400 to 1.4420 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text