Thermo Scientific Chemicals

2,4,6-Trichlorophenol, 98%, Thermo Scientific Chemicals

Catalog number: 139481000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4,6-Trichlorophenol, 98%, Thermo Scientific Chemicals

Catalog number: 139481000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 139481000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-06-2
IUPAC Name
2,4,6-trichlorophenol
Molecular Formula
C6H3Cl3O
InChI Key
LINPIYWFGCPVIE-UHFFFAOYSA-N
SMILES
OC1=C(Cl)C=C(Cl)C=C1Cl
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or mass and/or chunks
GC
>=97.5 %
Melting point
64°C to 68°C
Appearance (Color)
Light beige to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text