Triethanolamine, 97%, Thermo Scientific Chemicals
Triethanolamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethanolamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
2.5 L
25 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 139560010
Price (USD)/ Each
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS102-71-6
IUPAC Name2-[bis(2-hydroxyethyl)amino]ethan-1-ol
Molecular FormulaC6H15NO3
InChI KeyGSEJCLTVZPLZKY-UHFFFAOYSA-N
SMILESOCCN(CCO)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
Refractive index1.4830 to 1.4860 (20°C, 589 nm)
Appearance (Form)Clear viscous liquid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only