Thermo Scientific Chemicals

Triethyl borate, 97%, Thermo Scientific Chemicals

Catalog number: 139582500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Triethyl borate, 97%, Thermo Scientific Chemicals

Catalog number: 139582500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 139582500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
150-46-9
IUPAC Name
triethyl borate
Molecular Formula
C6H15BO3
InChI Key
AJSTXXYNEIHPMD-UHFFFAOYSA-N
SMILES
CCOB(OCC)OCC
Refractive index
1.3705 to 1.3775 (20°C, 589 nm)
Appearance (Color)
Clear colorless
GC
>=96.0 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text