1x1 image pixel for data collection
Triethylene glycol, 99%
Triethylene glycol, 99%
Triethylene glycol, 99%
Triethylene glycol, 99%
Thermo Scientific Chemicals

Triethylene glycol, 99%

CAS: 112-27-6 | C6H14O4 | 150.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 139590025
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS112-27-6
IUPAC Name2-[2-(2-hydroxyethoxy)ethoxy]ethan-1-ol
Molecular FormulaC6H14O4
InChI KeyZIBGPFATKBEMQZ-UHFFFAOYSA-N
SMILESOCCOCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Refractive index1.4545 to 1.4565 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link