Triethylene glycol dimethyl ether, 99%, stabilized, Thermo Scientific Chemicals
Triethylene glycol dimethyl ether, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethylene glycol dimethyl ether, 99%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass bottle
Catalog number 139600025
Price (USD)/ Each
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-49-2
IUPAC Name2,5,8,11-tetraoxadodecane
Molecular FormulaC8H18O4
InChI KeyYFNKIDBQEZZDLK-UHFFFAOYSA-N
SMILESCOCCOCCOCCOC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.4220 to 1.4240 (20°C, 589 nm)
Stabilizer0.01 % BHT
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only