Thermo Scientific Chemicals

Triethylene glycol dimethyl ether, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 139602500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Triethylene glycol dimethyl ether, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 139602500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 139602500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-49-2
IUPAC Name
2,5,8,11-tetraoxadodecane
Molecular Formula
C8H18O4
InChI Key
YFNKIDBQEZZDLK-UHFFFAOYSA-N
SMILES
COCCOCCOCCOC
Infrared spectrum
Conforms
Refractive index
1.4220 to 1.4240 (20°C, 589 nm)
Stabilizer
0.01 % BHT
GC
>=98.5 %
Water
=<0.1 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text