Thermo Scientific Chemicals

Triethyl 1,1,2-ethanetricarboxylate, 99%, Thermo Scientific Chemicals

Catalog number: 139611000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Triethyl 1,1,2-ethanetricarboxylate, 99%, Thermo Scientific Chemicals

Catalog number: 139611000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 139611000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7459-46-3
IUPAC Name
1,1,2-triethyl ethane-1,1,2-tricarboxylate
Molecular Formula
C11H18O6
InChI Key
TVWZLLYAJDSSCJ-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(C(=O)OCC)C(=O)OCC
Appearance (Color)
Clear colorless
GC
>=98.5 %
Infrared spectrum
Conforms
Refractive index
1.4280 to 1.4300 (20°C, 589 nm)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text