Thermo Scientific Chemicals

Triethyl methanetricarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: 139631000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Triethyl methanetricarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: 139631000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 139631000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6279-86-3
IUPAC Name
triethyl methanetricarboxylate
Molecular Formula
C10H16O6
InChI Key
AGZPNUZBDCYTBB-UHFFFAOYSA-N
SMILES
CCOC(=O)C(C(=O)OCC)C(=O)OCC
GC
>=97.5 %
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Appearance (Form)
Liquid after melting
Refractive index
1.4240 to 1.4260 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text