Triethyl orthopropionate, 98+%, Thermo Scientific Chemicals
Triethyl orthopropionate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethyl orthopropionate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
Catalog number 139670025
Price (USD)/ Each
463.65
Online exclusive
515.00 
Save 51.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)/ Each
463.65
Online exclusive
515.00 
Save 51.35 (10%)
Add to cart
Triethyl orthopropionate, 98+%, Thermo Scientific Chemicals
Catalog number139670025
Price (USD)/ Each
463.65
Online exclusive
515.00 
Save 51.35 (10%)
-
Add to cart
Chemical Identifiers
CAS115-80-0
IUPAC Name1,1,1-triethoxypropane
Molecular FormulaC9H20O3
InChI KeyFGWYWKIOMUZSQF-UHFFFAOYSA-N
SMILESCCOC(CC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=98.0 %
Infrared spectrumConforms
Refractive index1.4010 to 1.4030 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only