Thermo Scientific Chemicals

Triethyl orthopropionate, 98+%, Thermo Scientific Chemicals

Catalog number: 139671000
100 g, Each
Thermo Scientific Chemicals

Triethyl orthopropionate, 98+%, Thermo Scientific Chemicals

Catalog number: 139671000
100 g, Each
Quantity
Catalog number: 139671000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
115-80-0
IUPAC Name
1,1,1-triethoxypropane
Molecular Formula
C9H20O3
InChI Key
FGWYWKIOMUZSQF-UHFFFAOYSA-N
SMILES
CCOC(CC)(OCC)OCC
Appearance (Form)
Liquid
GC
>=98.0 %
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4010 to 1.4030 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text