Thermo Scientific Chemicals

Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals

Catalog number: 139705000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals

Catalog number: 139705000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 139705000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
867-13-0
IUPAC Name
ethyl 2-(diethoxyphosphoryl)acetate
Molecular Formula
C8H17O5P
InChI Key
GGUBFICZYGKNTD-UHFFFAOYSA-N
SMILES
CCOC(=O)CP(=O)(OCC)OCC
Color scale
=<50 APHA
Appearance (Form)
Clear liquid
Acid value
=<2 mg KOH/g
Infrared spectrum
Conforms
Refractive index
1.4300 to 1.4320 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text