Thermo Scientific Chemicals

Trifluoroacetic acid, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 139720025
2.5 kg, Glass Bottle, Each
Thermo Scientific Chemicals

Trifluoroacetic acid, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 139720025
2.5 kg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 139720025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-05-1
IUPAC Name
trifluoroacetic acid
Molecular Formula
C2HF3O2
InChI Key
DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES
OC(=O)C(F)(F)F
Infrared spectrum
Conforms
Water
=<0.3 % (Coulometric)
Appearance (Form)
Clear liquid
Titration with NaOH
99.0 to 101.0 %
Color scale
=<20 APHA

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text