Trifluoroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoroacetic acid, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass Bottle
Catalog number 139725000
Price (USD)/ Each
199.65
Special Offer
Online exclusive
Ends: 31-Mar-2025
249.00 
Save 49.35 (20%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
199.65
Special Offer
Online exclusive
Ends: 31-Mar-2025
249.00 
Save 49.35 (20%)
Add to cart
Trifluoroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
Catalog number139725000
Price (USD)/ Each
199.65
Special Offer
Online exclusive
Ends: 31-Mar-2025
249.00 
Save 49.35 (20%)
-
Add to cart
Chemical Identifiers
IUPAC Nametrifluoroacetic acid
Molecular FormulaC2HF3O2
InChI KeyDTQVDTLACAAQTR-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)F
Molecular Weight (g/mol)114.02
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.3 % (Coulometric)
Color scale=<20 APHA
Appearance (Form)Clear liquid
Titration with NaOH99.0 to 101.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only