Thermo Scientific Chemicals

Trifluoroacetic acid, silver salt, 98%, Thermo Scientific Chemicals

Catalog number: 139730050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Trifluoroacetic acid, silver salt, 98%, Thermo Scientific Chemicals

Catalog number: 139730050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 139730050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2966-50-9
IUPAC Name
silver(1+) trifluoroacetate
Molecular Formula
C2AgF3O2
InChI Key
KZJPVUDYAMEDRM-UHFFFAOYSA-M
SMILES
[Ag+].[O-]C(=O)C(F)(F)F
Appearance (Color)
Light beige to grey
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Titration with NH4SCN
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text