1,1,1-Trifluoroacetone, 98+%, Thermo Scientific Chemicals
1,1,1-Trifluoroacetone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1,1-Trifluoroacetone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 139740250
Price (USD)/ Each
121.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
121.00
Add to cart
1,1,1-Trifluoroacetone, 98+%, Thermo Scientific Chemicals
Catalog number139740250
Price (USD)/ Each
121.00
-
Add to cart
Chemical Identifiers
IUPAC Name1,1,1-trifluoropropan-2-one
Molecular FormulaC3H3F3O
InChI KeyFHUDAMLDXFJHJE-UHFFFAOYSA-N
SMILESCC(=O)C(F)(F)F
Molecular Weight (g/mol)112.05
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to orange
Infrared spectrumConforms
GC>=94.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only